Assay Details
| Gene Symbol: BGLU11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001123745.2 | NP_001117217.1 | 4 - 5 | 522 | 79 | |
| NM_001331360.1 | NP_001318909.1 | 4 - 5 | 522 | 79 | ||
| NM_100166.4 | NP_563666.1 | 4 - 5 | 522 | 79 | ||
| NM_179247.6 | NP_849578.5 | 4 - 5 | 522 | 79 | ||
| NM_202017.3 | NP_973746.3 | 4 - 5 | 391 | 79 | ||
| GenBank mRNA | AY049274.1 | - | 4 - 5 | 383 | 79 | |
| AY062763.1 | - | 4 - 5 | 384 | 79 | ||
| BT001137.1 | - | 4 - 5 | 327 | 79 | ||
More Information
Set Membership: |
|

BGLU11
Genomic Map