Assay Details
| Gene Symbol: ZIP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100409.5 | NP_172022.1 | 1 - 2 | 761 | 120 | |
| NM_202033.1 | NP_973762.1 | 1 - 2 | 710 | 120 | ||
| GenBank mRNA | AK176625.1 | - | 1 - 2 | 761 | 120 | |
| BT014902.1 | - | 1 - 2 | 694 | 120 | ||
More Information
Set Membership: |
|

ZIP5
Genomic Map