Assay Details
| Gene Symbol: CRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198007.1 | NP_001184936.1 | 10 - 11 | 1160 | 68 | |
| NM_100718.5 | NP_563816.1 | 11 - 12 | 1300 | 68 | ||
| NM_202064.1 | NP_973793.1 | 11 - 12 | 1006 | 68 | ||
| GenBank mRNA | AY056320.1 | - | 11 - 12 | 1168 | 68 | |
More Information
Set Membership: |
|

CRT3
Genomic Map