Assay Details
| Gene Symbol: CRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198007.1 | NP_001184936.1 | 6 - 7 | 875 | 71 | |
| NM_100718.5 | NP_563816.1 | 6 - 7 | 940 | 71 | ||
| NM_202064.1 | NP_973793.1 | 6 - 7 | 646 | 71 | ||
| GenBank mRNA | AY056320.1 | - | 6 - 7 | 808 | 71 | |
More Information
Set Membership: |
|

CRT3
Genomic Map