Assay Details
| Gene Symbol: CRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100718.5 | NP_563816.1 | 9 - 10 | 1153 | 152 | |
| NM_202064.1 | NP_973793.1 | 9 - 10 | 859 | 152 | ||
| GenBank mRNA | AY056320.1 | - | 9 - 10 | 1021 | 152 | |
More Information
Set Membership: |
|

CRT3
Genomic Map