Assay Details
| Gene Symbol: MYB60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331790.1 | NP_001318958.1 | 2 | 1024 | 164 | |
| NM_100755.3 | NP_172358.1 | 3 | 913 | 164 | ||
| NM_202066.1 | NP_973795.1 | 1 | 663 | 164 | ||
| GenBank mRNA | AK117469.1 | - | 3 | 783 | 164 | |
| AY519551.1 | - | 3 | 764 | 164 | ||
| BT005074.1 | - | 3 | 764 | 164 | ||
More Information
Set Membership: |
|

MYB60
Genomic Map