Assay Details
| Gene Symbol: NF-YC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100768.4 | NP_172371.1 | 2 | 948 | 116 | |
| GenBank mRNA | AY126997.1 | - | 2 | 915 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NF-YC9| Gene Symbol: NF-YC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100768.4 | NP_172371.1 | 2 | 948 | 116 | |
| GenBank mRNA | AY126997.1 | - | 2 | 915 | 116 | |
Set Membership: |
|