Assay Details
| Gene Symbol: NF-YC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179288.3 | NP_849619.1 | 2 - 3 | 906 | 74 | |
| NM_202068.2 | NP_973797.1 | 1 - 2 | 1308 | 74 | ||
| GenBank mRNA | AK316882.2 | - | 2 - 3 | 817 | 74 | |
More Information
Set Membership: |
|

NF-YC9
Genomic Map