Assay Details
| Gene Symbol: OTS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331913.1 | NP_001322371.1 | 5 - 6 | 1383 | 86 | |
| NM_100932.4 | NP_172527.2 | 6 - 7 | 1003 | 86 | ||
| NM_202073.4 | NP_973802.1 | 6 - 7 | 1187 | 86 | ||
| GenBank mRNA | AY092981.1 | - | 6 - 7 | 982 | 86 | |
| AY128798.1 | - | 6 - 7 | 937 | 86 | ||
More Information
Set Membership: |
|

OTS2
Genomic Map