Assay Details
| Gene Symbol: ADAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332205.1 | NP_001320850.1 | 2 - 3 | 537 | 61 | |
| NM_202110.2 | NP_973839.1 | 2 - 3 | 762 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001332205.1 transcripts.
Set Membership: |
|

ADAP
Genomic Map