Assay Details
| Gene Symbol: JAZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332386.1 | NP_001319041.1 | 3 | 1186 | 70 | |
| NM_101776.3 | NP_564075.1 | 2 | 996 | 70 | ||
| NM_202133.1 | NP_973862.1 | 1 | 1135 | 70 | ||
| GenBank mRNA | AY039894.1 | - | 2 | 933 | 70 | |
| AY065146.1 | - | 2 | 938 | 70 | ||
| AY081633.1 | - | 2 | 739 | 70 | ||
More Information
Set Membership: |
|

JAZ1
Genomic Map