Assay Details
| Gene Symbol: AT1G21610 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332521.1 | NP_001321246.1 | 11 - 12 | 2307 | 84 | |
| NM_001332522.1 | NP_001321247.1 | 11 - 12 | 2181 | 84 | ||
| NM_001332523.1 | NP_001321248.1 | 12 - 13 | 2026 | 84 | ||
| NM_102010.5 | NP_173580.2 | 13 - 14 | 1862 | 84 | ||
| NM_202151.3 | NP_973880.1 | 13 - 14 | 1934 | 84 | ||
| GenBank mRNA | AY090255.1 | - | 13 - 14 | 1818 | 84 | |
More Information
Set Membership: |
|

AT1G21610
Genomic Map