Assay Details
| Gene Symbol: GRF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102081.4 | NP_564167.1 | 2 - 3 | 668 | 69 | |
| NM_179367.2 | NP_849698.1 | 2 - 3 | 492 | 69 | ||
| NM_202155.2 | NP_973884.1 | 2 - 3 | 492 | 69 | ||
| GenBank mRNA | AF334382.1 | - | 2 - 3 | 376 | 69 | |
| AK316870.1 | - | 2 - 3 | 423 | 69 | ||
| AY054505.1 | - | 2 - 3 | 423 | 69 | ||
| AY058834.1 | - | 2 - 3 | 475 | 69 | ||
| AY062838.1 | - | 2 - 3 | 423 | 69 | ||
| AY081674.1 | - | 2 - 3 | 376 | 69 | ||
| AY087580.1 | - | 2 - 3 | 469 | 69 | ||
| U36446.1 | - | 2 - 3 | 424 | 69 | ||
More Information
Set Membership: |
|

GRF10
Genomic Map