Assay Details
| Gene Symbol: MENG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102185.4 | NP_173750.3 | 5 - 6 | 576 | 87 | |
| NM_202165.1 | NP_973894.1 | 2 - 3 | 809 | 87 | ||
| NM_202166.2 | NP_973895.1 | 4 - 5 | 652 | 87 | ||
| GenBank mRNA | AK317221.1 | - | 5 - 6 | 555 | 87 | |
| BT014858.1 | - | 2 - 3 | 225 | 87 | ||
More Information
Set Membership: |
|

MENG
Genomic Map