Assay Details
| Gene Symbol: MENG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_102185.4 | NP_173750.3 | 7 - 8 | 731 | 106 | |
| NM_202165.1 | NP_973894.1 | 4 - 5 | 964 | 106 | ||
| NM_202166.2 | NP_973895.1 | 6 - 7 | 807 | 106 | ||
| GenBank mRNA | AK317221.1 | - | 7 - 8 | 710 | 106 | |
| BT014858.1 | - | 4 - 5 | 380 | 106 | ||
More Information
Set Membership: |
|

MENG
Genomic Map