Assay Details
| Gene Symbol: AT1G27040 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332738.1 | NP_001322135.1 | 2 - 3 | 777 | 134 | |
| NM_102466.4 | NP_174024.2 | 4 - 5 | 797 | 134 | ||
| NM_202190.1 | NP_973919.1 | 3 - 4 | 837 | 134 | ||
| GenBank mRNA | AY072134.1 | - | 4 - 5 | 797 | 134 | |
| AY096445.1 | - | 3 - 4 | 773 | 134 | ||
More Information
Set Membership: |
|

AT1G27040
Genomic Map