Assay Details
| Gene Symbol: AT3G04480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111319.4 | NP_187098.2 | 5 - 6 | 592 | 118 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT3G04480| Gene Symbol: AT3G04480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111319.4 | NP_187098.2 | 5 - 6 | 592 | 118 | |
Set Membership: |
|