Assay Details
| Gene Symbol: THA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202878.1 | NP_001189807.1 | 5 - 6 | 788 | 69 | |
| NM_001337534.1 | NP_001327633.1 | 4 - 5 | 798 | 69 | ||
| NM_111323.4 | NP_566228.1 | 5 - 6 | 864 | 69 | ||
| GenBank mRNA | AF325033.2 | - | 5 - 6 | 752 | 69 | |
| AK175345.1 | - | 5 - 6 | 724 | 69 | ||
| AY085456.1 | - | 5 - 6 | 724 | 69 | ||
| AY091093.1 | - | 5 - 6 | 776 | 69 | ||
| AY123033.1 | - | 4 - 5 | 686 | 69 | ||
More Information
Set Membership: |
|

THA2
Genomic Map