Assay Details
| Gene Symbol: THA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202878.1 | NP_001189807.1 | 8 - 9 | 1077 | 70 | |
| NM_001337534.1 | NP_001327633.1 | 7 - 8 | 1090 | 70 | ||
| NM_111323.4 | NP_566228.1 | 8 - 9 | 1156 | 70 | ||
| GenBank mRNA | AF325033.2 | - | 8 - 9 | 1044 | 70 | |
| AK175345.1 | - | 8 - 9 | 1016 | 70 | ||
| AY085456.1 | - | 8 - 9 | 1016 | 70 | ||
| AY091093.1 | - | 8 - 9 | 1068 | 70 | ||
| AY123033.1 | - | 7 - 8 | 978 | 70 | ||
More Information
Set Membership: |
|

THA2
Genomic Map