Assay Details
| Gene Symbol: THA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202878.1 | NP_001189807.1 | 9 - 10 | 1143 | 110 | |
| NM_001337534.1 | NP_001327633.1 | 8 - 9 | 1156 | 110 | ||
| NM_111323.4 | NP_566228.1 | 9 - 10 | 1222 | 110 | ||
| GenBank mRNA | AF325033.2 | - | 9 - 10 | 1110 | 110 | |
| AK175345.1 | - | 9 - 10 | 1082 | 110 | ||
| AY085456.1 | - | 9 - 10 | 1082 | 110 | ||
| AY091093.1 | - | 9 - 10 | 1134 | 110 | ||
More Information
Set Membership: |
|

THA2
Genomic Map