Assay Details
| Gene Symbol: CLPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337539.1 | NP_001319471.1 | 5 - 6 | 894 | 76 | |
| NM_001337540.1 | NP_001327497.1 | 4 - 5 | 1142 | 76 | ||
| NM_111340.3 | NP_187119.1 | 5 - 6 | 931 | 76 | ||
| GenBank mRNA | AY062866.1 | - | 5 - 6 | 889 | 76 | |
| AY128751.1 | - | 4 - 5 | 623 | 76 | ||
More Information
Set Membership: |
|

CLPS3
Genomic Map