Assay Details
| Gene Symbol: CLPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337539.1 | NP_001319471.1 | 7 - 8 | 1226 | 84 | |
| NM_001337540.1 | NP_001327497.1 | 6 - 7 | 1474 | 84 | ||
| NM_111340.3 | NP_187119.1 | 7 - 8 | 1263 | 84 | ||
| GenBank mRNA | AY062866.1 | - | 7 - 8 | 1221 | 84 | |
| AY128751.1 | - | 6 - 7 | 955 | 84 | ||
More Information
Set Membership: |
|

CLPS3
Genomic Map