Assay Details
| Gene Symbol: TPR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035558.1 | NP_001030635.1 | 11 - 12 | 1372 | 90 | |
| NM_001202879.1 | NP_001189808.1 | 13 - 14 | 2182 | 90 | ||
| NM_111343.3 | NP_187122.1 | 11 - 12 | 1431 | 90 | ||
| GenBank mRNA | AK175477.1 | - | 11 - 12 | 1375 | 90 | |
| AY084331.1 | - | 11 - 12 | 1379 | 90 | ||
| AY136361.1 | - | 11 - 12 | 1379 | 90 | ||
| BT000201.1 | - | 11 - 12 | 1230 | 90 | ||
More Information
Set Membership: |
|

TPR10
Genomic Map