Assay Details
| Gene Symbol: AT3G04850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111357.4 | NP_187136.3 | 8 - 9 | 1967 | 99 | |
| GenBank mRNA | BT003926.1 | - | 8 - 9 | 1884 | 99 | |
| BT005010.1 | - | 8 - 9 | 1742 | 99 | ||
More Information
Set Membership: |
|

AT3G04850
Genomic Map