Assay Details
| Gene Symbol: AT3G04850 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111357.4 | NP_187136.3 | 9 - 10 | 2038 | 107 | |
| GenBank mRNA | BT003926.1 | - | 9 - 10 | 1955 | 107 | |
| BT005010.1 | - | 9 - 10 | 1813 | 107 | ||
More Information
Set Membership: |
|

AT3G04850
Genomic Map