Assay Details
| Gene Symbol: CYSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337549.1 | NP_001325893.1 | 2 - 3 | 353 | 102 | |
| NM_111366.4 | NP_566243.1 | 3 - 4 | 368 | 102 | ||
| GenBank mRNA | AB024284.1 | - | 3 - 4 | 278 | 102 | |
| AJ011603.1 | - | 3 - 4 | 278 | 102 | ||
| AY136420.1 | - | 3 - 4 | 242 | 102 | ||
| BT008721.1 | - | 2 - 3 | 150 | 102 | ||
More Information
Set Membership: |
|

CYSD1
Genomic Map