Assay Details
| Gene Symbol: CYSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337549.1 | NP_001325893.1 | 6 - 7 | 869 | 77 | |
| NM_111366.4 | NP_566243.1 | 7 - 8 | 884 | 77 | ||
| GenBank mRNA | AB024284.1 | - | 7 - 8 | 794 | 77 | |
| AJ011603.1 | - | 7 - 8 | 794 | 77 | ||
| AY136420.1 | - | 7 - 8 | 758 | 77 | ||
| BT008721.1 | - | 6 - 7 | 666 | 77 | ||
More Information
Set Membership: |
|

CYSD1
Genomic Map