Assay Details
| Gene Symbol: ACP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111374.2 | NP_187153.1 | 1 - 2 | 128 | 64 | |
| GenBank mRNA | AK227721.1 | - | 1 - 2 | 128 | 64 | |
| AY085288.1 | - | 1 - 2 | 129 | 64 | ||
| BT006397.1 | - | 1 - 2 | 52 | 64 | ||
More Information
Set Membership: |
|

ACP1
Genomic Map