Assay Details
| Gene Symbol: MIRO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337575.1 | NP_001327041.1 | 3 - 4 | 690 | 155 | |
| NM_001337576.1 | NP_001327040.1 | 3 - 4 | 690 | 155 | ||
| NM_111404.1 | NP_187182.1 | 5 - 6 | 926 | 155 | ||
More Information
Set Membership: |
|

MIRO3
Genomic Map