Assay Details
| Gene Symbol: MIRO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337575.1 | NP_001327041.1 | 4 - 5 | 764 | 143 | |
| NM_001337576.1 | NP_001327040.1 | 4 - 5 | 764 | 143 | ||
| NM_111404.1 | NP_187182.1 | 6 - 7 | 1000 | 143 | ||
More Information
Set Membership: |
|

MIRO3
Genomic Map