Assay Details
| Gene Symbol: ALY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035566.2 | NP_001030643.1 | 18 - 19 | 3190 | 58 | |
| NM_001125108.2 | NP_001118580.1 | 17 - 18 | 3276 | 58 | ||
| NM_001202883.1 | NP_001189812.1 | 18 - 19 | 3155 | 58 | ||
| NM_001202884.1 | NP_001189813.1 | 19 - 20 | 3123 | 58 | ||
| NM_111411.5 | NP_187189.2 | 18 - 19 | 3187 | 58 | ||
| GenBank mRNA | AJ583496.1 | - | 18 - 19 | 3127 | 58 | |
More Information
Set Membership: |
|

ALY2
Genomic Map