Assay Details
| Gene Symbol: ALY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035566.2 | NP_001030643.1 | 3 - 4 | 391 | 59 | |
| NM_001125108.2 | NP_001118580.1 | 2 - 3 | 477 | 59 | ||
| NM_001202883.1 | NP_001189812.1 | 3 - 4 | 356 | 59 | ||
| NM_001202884.1 | NP_001189813.1 | 4 - 5 | 324 | 59 | ||
| NM_111411.5 | NP_187189.2 | 3 - 4 | 388 | 59 | ||
| GenBank mRNA | AJ583496.1 | - | 3 - 4 | 328 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001125108.2 transcripts.
Set Membership: |
|

ALY2
Genomic Map