Assay Details
| Gene Symbol: RPT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111426.4 | NP_187204.1 | 9 - 10 | 1382 | 73 | |
| GenBank mRNA | AY062705.1 | - | 9 - 10 | 1308 | 73 | |
More Information
Set Membership: |
|

RPT5A
Genomic Map