Assay Details
| Gene Symbol: ATCHR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111475.3 | NP_187252.2 | 11 - 12 | 2867 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATCHR12| Gene Symbol: ATCHR12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111475.3 | NP_187252.2 | 11 - 12 | 2867 | 84 | |
Set Membership: |
|