Assay Details
| Gene Symbol: FRS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111500.3 | NP_566278.1 | 2 - 3 | 2672 | 77 | |
| GenBank mRNA | AF360270.1 | - | 2 - 3 | 2617 | 77 | |
More Information
Set Membership: |
|

FRS7
Genomic Map