Assay Details
Gene Symbol: SFR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125119.2 | NP_001118591.1 | 4 - 5 | 867 | 139 | |
NM_111527.3 | NP_566285.1 | 4 - 5 | 950 | 139 | ||
GenBank mRNA | AJ491320.1 | - | 4 - 5 | 866 | 139 | |
AY059889.1 | - | 4 - 5 | 791 | 139 | ||
BT002100.1 | - | 4 - 5 | 746 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
