Assay Details
| Gene Symbol: SFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125119.2 | NP_001118591.1 | 5 - 6 | 1086 | 68 | |
| NM_111527.3 | NP_566285.1 | 5 - 6 | 1169 | 68 | ||
| GenBank mRNA | AJ491320.1 | - | 5 - 6 | 1085 | 68 | |
| AY059889.1 | - | 5 - 6 | 1010 | 68 | ||
| BT002100.1 | - | 5 - 6 | 965 | 68 | ||
More Information
Set Membership: |
|

SFR2
Genomic Map