Assay Details
| Gene Symbol: AGL91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111544.1 | NP_187320.1 | 1 | 467 | 95 | |
| GenBank mRNA | DQ056634.1 | - | 1 | 467 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AGL91| Gene Symbol: AGL91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111544.1 | NP_187320.1 | 1 | 467 | 95 | |
| GenBank mRNA | DQ056634.1 | - | 1 | 467 | 95 | |
Set Membership: |
|