Assay Details
| Gene Symbol: ALDH22A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111545.3 | NP_187321.2 | 2 - 3 | 285 | 69 | |
| NM_202513.3 | NP_974242.1 | 2 - 3 | 398 | 69 | ||
| GenBank mRNA | AJ584646.1 | - | 2 - 3 | 168 | 69 | |
More Information
Set Membership: |
|

ALDH22A1
Genomic Map