Assay Details
| Gene Symbol: TRX z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111553.4 | NP_187329.1 | 1 - 2 | 349 | 82 | |
| GenBank mRNA | AF370315.1 | - | 1 - 2 | 245 | 82 | |
| AY063100.1 | - | 1 - 2 | 228 | 82 | ||
More Information
Set Membership: |
|

TRX z
Genomic Map