Assay Details
| Gene Symbol: AT3G06980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111578.2 | NP_187354.1 | 2 - 3 | 1307 | 95 | |
| GenBank mRNA | AJ296275.1 | - | 2 - 3 | 1262 | 95 | |
| AY048260.1 | - | 2 - 3 | 1272 | 95 | ||
| AY113064.1 | - | 2 - 3 | 1208 | 95 | ||
More Information
Set Membership: |
|

AT3G06980
Genomic Map