Assay Details
| Gene Symbol: AT3G06980 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111578.2 | NP_187354.1 | 7 - 8 | 2212 | 83 | |
| GenBank mRNA | AJ296275.1 | - | 7 - 8 | 2167 | 83 | |
| AY048260.1 | - | 7 - 8 | 2177 | 83 | ||
| AY113064.1 | - | 7 - 8 | 2113 | 83 | ||
More Information
Set Membership: |
|

AT3G06980
Genomic Map