Assay Details
| Gene Symbol: NS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337712.1 | NP_001326578.1 | 5 - 6 | 2069 | 58 | |
| NM_111621.4 | NP_187398.1 | 6 - 7 | 2011 | 58 | ||
More Information
Set Membership: |
|

NS2
Genomic Map