Assay Details
| Gene Symbol: TK1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111659.4 | NP_187437.1 | 1 - 2 | 428 | 84 | |
| GenBank mRNA | AF370344.1 | - | 1 - 2 | 259 | 84 | |
| AY142670.1 | - | 1 - 2 | 183 | 84 | ||
More Information
Set Membership: |
|

TK1a
Genomic Map