Assay Details
| Gene Symbol: PLC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035583.1 | NP_001030660.1 | 1 - 2 | 596 | 120 | |
| NM_001161124.1 | NP_001154596.1 | 1 - 2 | 596 | 120 | ||
| NM_001337760.1 | NP_001327169.1 | 1 - 2 | 596 | 120 | ||
| NM_111686.6 | NP_187464.1 | 1 - 2 | 643 | 120 | ||
| GenBank mRNA | AF360206.1 | - | 1 - 2 | 364 | 120 | |
| AK318857.1 | - | 1 - 2 | 454 | 120 | ||
| AY040054.1 | - | 1 - 2 | 301 | 120 | ||
| D50804.1 | - | 1 - 2 | 475 | 120 | ||
More Information
Set Membership: |
|

PLC2
Genomic Map