Assay Details
| Gene Symbol: UGE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118524.3 | NP_194123.1 | 8 - 9 | 1271 | 85 | |
| GenBank mRNA | AK118722.1 | - | 8 - 9 | 1102 | 85 | |
| BT008539.1 | - | 8 - 9 | 952 | 85 | ||
More Information
Set Membership: |
|

UGE2
Genomic Map