Assay Details
| Gene Symbol: ALL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341640.1 | NP_001328202.1 | 1 - 2 | 637 | 117 | |
| NM_118540.3 | NP_194139.2 | 2 - 3 | 382 | 117 | ||
More Information
Set Membership: |
|

ALL1
Genomic Map