Assay Details
| Gene Symbol: EMB140 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001341660.1 | NP_001328782.1 | 12 - 13 | 1742 | 99 | |
| NM_001341661.1 | NP_001328783.1 | 12 - 13 | 1751 | 99 | ||
| NM_118560.4 | NP_194158.3 | 12 - 13 | 1816 | 99 | ||
| NM_179220.2 | NP_849551.1 | 12 - 13 | 1751 | 99 | ||
More Information
Set Membership: |
|

EMB140
Genomic Map