Assay Details
| Gene Symbol: ATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118585.4 | NP_194183.1 | 1 - 2 | 497 | 111 | |
| GenBank mRNA | AY054688.1 | - | 1 - 2 | 390 | 111 | |
| BT008426.1 | - | 1 - 2 | 301 | 111 | ||
More Information
Set Membership: |
|

ATR1
Genomic Map