Assay Details
| Gene Symbol: ATR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203894.1 | NP_001190823.1 | 6 - 7 | 745 | 122 | |
| NM_118585.4 | NP_194183.1 | 6 - 7 | 820 | 122 | ||
| GenBank mRNA | AY054688.1 | - | 6 - 7 | 713 | 122 | |
| BT008426.1 | - | 6 - 7 | 624 | 122 | ||
More Information
Set Membership: |
|

ATR1
Genomic Map